Learn More
Hecht Karl™ Class B DURAN Borosilicate Glass Tall Form Measuring Cylinders
Specifica
Specifica
| Tolleranza | ±15 mL |
| Materiale | Borosilicate Glass |
| Scale Color | Blue |
| Certificazione/Conformità | DIN EN ISO 4788 |
| Classe | B |
| Capacità (sistema metrico) | 2000 mL |
| Bilancia | Blue Graduations |
| Tolerance (Metric) | ±15mL |
| Product Type | Graduated Measuring Cylinder |
| Limite di errore | ±15mL |