Learn More
Fisherbrand Class II Biological Safety Cabinet
Codice prodotto. 30564668 Sfoglia Tutto Fisherbrand Traceable Prodotti
Cambia vista
Click to view available options
Lunghezza (sistema metrico) parte esterna:
1.2 m
1.8 m
Tipo di spina:
Schuko
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Lunghezza (sistema metrico) parte esterna Tipo di spina Conditionnement
30564668 1.8 m Schuko Pezzo
30564665 1.2 m Schuko Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 30564668
Equipment and Instrument End of Year Sale
Usa il codice promozionale 28000
Afficher toutes les promotions
 Articolo soggetto a restrizioni
In stock
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Fisherbrand Class II Biological Safety Cabinet is an economical solution for high protection.
  • Fisherbrand Class II biological safety cabinet (BSC) provides both personal and product protection from biological hazards and contaminations.
  • This model is available in 2 sizes; 1.2 m and 1.8 m with 3 different plug and socket options (Type F, Type G, and Type E)
  • Equipped with technology to automate airflow compensation and alerts user to out of tolerance airflows that could compromise product and operator protections
  • H14 HEPA filters removes airborne contaminants
  • 10-degrees sloped front for exceptional comfort and ergonomics
  • SmartClean front window design to improve access to the work area for cleaning
  • Night Set Back Mode reduces airflow during low-usage periods
  • The BSC's comply with all applicable performance and safety requirements of EN12469:2000
TRUSTED_SUSTAINABILITY
Spécification
Certificazione/Conformità EN 12469
Materiale camera Stainless Steel (304)
Product Type Class II Biosafety Cabinet
Modello Fisherbrand BSC 1.8
Livello di rumorosità <60 dBA
Prese 2, 2 Single, Rear Wall
Tipo di spina Schuko
Consumo energetico 2000 W
Filtro dell'aria di scarico HEPA H14 EN1822
Efficienza filtro 99.995% at MPPS
Include UV light, armrest and adjustable floor stand
Assorbimento elettrico (modalità flusso ridotto) 120 W
Lighting LED
Porte 2 x 2
Filtro dell'aria in entrata HEPA H14 EN1822
Warranty 2 Years
Area della superficie di lavoro 120 x 45 cm
Dimensioni esterne (L x P x A) 130 x 80.2 x 152 cm
Lunghezza (sistema metrico) parte esterna 1.8 m
Depth (Metric) Interior 63 cm
Requisiti elettrici 230 V, 50/60 Hz
Tensione 230 V
Assorbimento elettrico (punto di regolazione operativo) 400 W
Quantità 1 Ea.
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.