Learn More
Abeba™ Classic 31010 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Marca:  Abeba™ 31010/44
Dettagli aggiuntivi : Peso : 0.95000kg
Codice prodotto. 15618287
  • 57.85€ / 1 paio
Spedizione stimata
18-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 SB SRC
  • Grained leather black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Heel patch microfiber
Specifica
Specifica
Animal fats, oil and fuel
Black
Leather
Slip-resistant
31010
CE EN ISO 20345:2011 SB SRC
Microfiber
44
Polyurethane
Steel