Learn More
Nickel Electro™ Clifton™ Centrifuge Rotor Angle Head
Nickel Electro™ Clifton™ Centrifuge Rotor Angle Head comes in different capacities that hold either 8, 12, or 24 tubes.
dietro preventivo
Specifica
Da utilizzare con (apparecchiatura) | Universal Rotor |
---|
Includes: dimCapacity
Descrizione
- Can be removed from centrifuge for cleaning
- The advantage of the angle head is that tubes which have large screw caps can be centrifuged.
- The large capped tubes obstruct the normal swing out head.
- Converts normal swing out head range Centrifuge to a fixed head.
Universal rotor, angle head, fixed head
Specifica
Universal Rotor |