Learn More
Jenway™ PFP7/C Clinical Flame Photometer
Designed for use in clinical applications for the determination of sodium, potassium and lithium. Jenway™ PFP7/C Clinical Flame Photometer features built-in linearizer circuitry that allows readings of both sodium and potassium to be displayed directly in mmol/l.
Marca:  Jenway™ 12655-00
Codice prodotto. 11495059
  • ONLINE EXCLUSIVE PRICE
    6585.00€
    5395.14€ / pezzi 1
    Risparmia 1189.86€
    18% Sconto
Spedizione stimata: 02-02-2027
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Designed for clinical analysis
  • Supplied with Na, K, Li, Ba and Ca filters
  • Low temperature, single channel
  • Flame failure safety system
  • Operates with propane, butane, natural gas or LPG
  • 3 year warranty
Specifica
Specifica
Clinical Flame Photometer PFP7