missing translation for 'onlineSavingsMsg'
Learn More

Clobetasone 17-Propionate, TRC

Codice prodotto. 30353010
Cambia vista
Click to view available options
Quantità:
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30353010 10 mg -
30359827 50 mg -
30361536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30353010 Supplier Toronto Research Chemicals Supplier No. TRCC58353510MG
 Articolo soggetto a restrizioni
Spedizione stimata: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25122-56-9
Formula molecolare C25 H30 Cl F O5
Molecular Weight (g/mol) 464.95
Sinonimo Clobetasone 17-Propionate, (16β)-21-Chloro-9-fluoro-16-methyl-3,11,20-trioxopregna-1,4-dien-17-yl Propionate, (16β)-21-Chloro-9-fluoro-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,11,20-trione, 21-Chloro-9-fluoro-17-hydroxy-16β-methylpregna-1,4-diene-3,11,20-trione propionate, Clobetasone propionate, Clobetasone-17-propionate
IUPAC Name [(8S,9R,10S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-10,13,16-trimethyl-3,11-dioxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] propanoate
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl

Specifications

Materiale o nome chimico Clobetasone 17-Propionate
Note sul grado di purezza HPLC
Quantità 10 mg
InChI Formula InChI=1S/C25H30ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-18H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,22-,23-,24-,25-/m0/s1
Peso formulazione 464.1766
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.