missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cloxacillin sodium monohydrate, Dr. Ehrenstorfer
Specifications
Specifications
| Materiale o nome chimico | Cloxacillin Sodium Monohydrate |
| CAS | 7081-44-9 |
| Nomi componenti o analita | Cloxacillin Sodium Monohydrate |
| Shipping Condition | +20°C |
| Formula molecolare | C19 H17 Cl N3 O5 S . Na . H2 O |
| Certificazione/Conformità | ISO 17034 |
| Sinonimo | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[3-(o-chlorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-, monosodium salt, monohydrate (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, monohydrate, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chlorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, monohydrate, [2S-(2α,5α,6β)]-, 3-(o-Chlorophenyl)-5-methyl-4-isoxazolyl penicillin sodium salt monohydrate, Cloxacillin sodium hydrate, Sodium 6-[3-(o-Chlorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate, Sodium cloxacillin monohydrate |
| InChI Formula | InChI=1S/C19H18ClN3O5S.Na.H2O/c1-8-11(12(22-28-8)9-6-4-5-7-10(9)20)15(24)21-13-16(25)23-14(18(26)27)19(2,3)29-17(13)23;;/h4-7,13-14,17H,1-3H3,(H,21,24)(H,26,27);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1 |
| SMILES | O.[Na+].Cc1onc(c2ccccc2Cl)c1C(=O)N[C@H]3[C@H]4SC(C)(C)[C@@H](N4C3=O)C(=O)[O-] |
| IUPAC Name | sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?