Descrizione
- Essential wrap-around comfort
- Upper protection
- Panoramic vision
- Non-slip bridge
- Comfortable, non-slip straight temples
- Temple or strap version with interchangeable foam
Specifica
Specifica
| Certificazione/Conformità | Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T; Solar radiations risks: EN 172 |
| Lens Coating | Anti-fog, Scratch-resistant |
| Descrizione | Tinted; Low energy impact, resists a 6 mm, 0.86g ball at 45 m/s |
| Lens Tint | Smoke |
| Colore telaio | Black |
| Stile stanghette | Interchangeable |
| Peso (sistema metrico) | 25 g |