missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme A Trilithium Salt Dihydrate, 96%, Fisher BioReagents

Catalog Number p-3200757
Click to view available options
Quantità:
50 mg
250 mg
1 g
Imballaggio:
Amber Glass
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
18439-24-2
C21H33Li3N7O16P3S
785.329
QSCBPHBAFBVXRK-UHFFFAOYSA-K
coenzyme a, trilithium salt
53442190
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
18439-24-2
C21H33Li3N7O16P3S
785.329
QSCBPHBAFBVXRK-UHFFFAOYSA-K
coenzyme a, trilithium salt
53442190
trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
[Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
Materiale o nome chimico Coenzyme A
Colore White
Intervallo percentuale saggio ≥96 %
Imballaggio Amber Glass
Quantità 50 mg
Percent Purity 96% min.
Forma fisica Powder or Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'