missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen X Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549198
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Collagen X Polyclonal antibody specifically detects Collagen X in Human samples. It is validated for Western Blot.Specifica
Collagen X | |
Polyclonal | |
Unconjugated | |
COL10A1 | |
Col10; COL10A1; Col10a-1; Collagen alpha-1(X) chain; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1300 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
Q03692 | |
COL10A1 | |
KLH conjugated synthetic peptide between 29-58 amino acids from the N-terminal region of human COL10A1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |