missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL15A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551288
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total COL15A1 protein.
COL15A1 Polyclonal antibody specifically detects COL15A1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifica
COL15A1 | |
Polyclonal | |
Unconjugated | |
COL15A1 | |
COFA1; COL15A1; Collagen alpha-1(XV) chain; collagen type XV alpha 1 chain; collagen type XV proteoglycan; collagen XV, alpha-1 polypeptide; collagen, type XV, alpha 1; endostatin-XV; procollagen type XV; procollagen, type XV; Related to endostatin; Restin; Restin-2; Restin-3; Restin-4; Restin-I; Restin-II; Restin-III; Restin-IV; type XV collagen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1306 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P39059 | |
COL15A1 | |
Synthetic peptide of human COL15A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |