Learn More
COL15A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553668
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MPGEVEASGV APGELDLSMS AQSLGEEATV GPSSEDSLTT AAAATEVSLS TFEDEEASGV PTDGLAPLTA TMAPERAVTS GPGDEEDLAA A Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 33%.
COL15A1 Polyclonal antibody specifically detects COL15A1 in Human samples. It is validated for Immunohistochemistry.Specifica
COL15A1 | |
Polyclonal | |
Unconjugated | |
COL15A1 | |
COFA1; COL15A1; Collagen alpha-1(XV) chain; collagen type XV alpha 1 chain; collagen type XV proteoglycan; collagen XV, alpha-1 polypeptide; collagen, type XV, alpha 1; endostatin-XV; procollagen type XV; procollagen, type XV; Related to endostatin; Restin; Restin-2; Restin-3; Restin-4; Restin-I; Restin-II; Restin-III; Restin-IV; type XV collagen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1306 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P39059 | |
COL15A1 | |
Recombinant protein corresponding to Human COL15A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only