missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL1A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550938
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
COL1A2 Polyclonal Antibody detects endogenous levels of COL1A2 protein.
COL1A2 Polyclonal antibody specifically detects COL1A2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifica
COL1A2 | |
Polyclonal | |
Unconjugated | |
COL1A2 | |
AA960264; AI325291; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; COL1; COL1A2; Col1a-2; COLA; Cola2; Cola-2; collagen; collagen a2; collagen alpha 2(I) chain precursor; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 2 chain; collagen, type I, alpha 2; OI4; oim; osteogenesis imperfecta; precollagen alpha-2; procollagen, type I, alpha 2; type I collagen alpha-2 chain; type I procollagen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1278, 12843, 84352 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P02466, P08123, Q01149 | |
COL1A2 | |
Synthesized peptide derived from the N-terminal region of human COL1A2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |