missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL24A1 Monoclonal Antibody (1D8), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524533
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Peptide Sequence: PRWTSTQTSG PGLPIGFKGW NGQIFKVNTL LEPKVLSDDC KIQDGSWHKA TFLFHTQEPN QLPVIEVQKL PHLKTERKYY IDSSSVCFL
COL24A1 Monoclonal antibody specifically detects COL24A1 in Human samples. It is validated for ELISA, Western Blot.Specifica
COL24A1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q17RW2 | |
COL24A1 | |
COL24A1 (NP_690850, 1626 a.a. ∽ 1714 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
1D8 | |
Unconjugated | |
COL24A1 | |
COL24A1; Collagen alpha-1(XXIV) chain; collagen type XXIV alpha 1; collagen, type XXIV, alpha 1 | |
Mouse | |
Protein A | |
RUO | |
255631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |