missing translation for 'onlineSavingsMsg'
Learn More

Abcam Collagen I, Native Bovine, Full Length, FITC, 1 mg

Codice prodotto. 32140746
Cambia vista
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
32140746 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32140746 Supplier Abcam Supplier No. ab3142011mg
 Articolo soggetto a restrizioni
Spedizione stimata: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FITC-conjugated native bovine collagen protein for research applications.

  • Protein Type: Native
  • Source: Cow (Bovine)
  • Conjugation: FITC
  • Form: Lyophilized; Constituents: 100% Tris buffered saline
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Tampone Tris buffered saline
Concentrazione 5 mg/mL
Content And Storage Store at -20°C.
Forma Lyophilized
Label or Dye FITC
Shipping Condition Blue Ice
Target Molecule Collagen I (Bovine)
Metodo di rilevamento Fluorescence
Formulazione Lyophilized from 100% Tris buffered saline
Quantità 1 mg
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.