Learn More
SymLine Collector, V2.0, 3/4-ways
Product Code. 17148698
Change view
Click to view available options
For Use With (Equipment):
SymLine System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17148698 SymLine System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17148698 Supplier SymLine Supplier No. 306509
  • ONLINE EXCLUSIVE PRICE
    254.00€
    232.44€ / pezzi 1
    Risparmia 21.56€
    8% Sconto
Spedizione stimata: 10-12-2026
This purging manifold allows you to install up to four SafetyCaps on your purging bottle.
  • Safe and easy handling - Capillaries are reliably fixed in place
  • Solvent filters have sufficient space due to the wide GL45 opening
  • Unused connections can be easily closed with the supplied blank cap
The system for Solvent Safety remains permanently closed, the safety chain remains intact during the rinsing process.
Specifications
For Use With (Application) Waste Disposal System
Description Collector, V2.0, universal 3/4-ways incl. blind cap for HPLC connection, 1 x GL45 (f) to 4 x GL45 (m), PE-HD electrostatic conductive
For Use With (Equipment) SymLine System
Type Replacement Screw Cap
Solvent Channels 4
Material HDPE-EC
Includes Universal 3/4-ways incl. blind cap for HPLC connection, 1 x GL45 (f) to 4 x GL45 (m)
Thread Size GL45