missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ 3´ mRNA Library Prep Kit for Illumina™ Systems

Designed for robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) platforms.

Marca:  Invitrogen™ A38110096

Dettagli aggiuntivi : Peso : 0.20000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17113149

  • 2470.00€ / 96 reazioni

In stock
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Supports library preparation from polyadenylated RNA in ∼4.5 hours.
  • The protocol generates one fragment per transcript, resulting in accurate gene expression values.
  • Sequences obtained are close to the 3’ ends of the transcripts, so the kit is suitable for varying-quality
  • RNA samples, including formalin-fixed paraffin-embedded (FFPE) samples.
  • The majority of inserts are greater than 75 bp in size, corresponding to final library fragment sizes ≥200 bp.
  • Multiplexing of libraries can be carried out using up to 96 single-indexed primers.
  • For convenience, the Collibri 3’ mRNA Library Prep Kit provides color-coded components for visual tracking of library preparation progress.
  • Inert dyes in the reagents provide visual confirmation of additions and mixing, but do not interfere with enzymatic reactions and do not compromise library prep and sequencing results.

Specifica

Specifica

• Collibri Library Prep Kit, store at -20°C
• Collibri Library Cleanup Kit, store at 4°C
Transcriptome Sequencing
Sequencing
96
mRNA Library Prep Kit
96 reactions
Kit
HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
cDNA Library
Collibri™
Approved for shipment on Wet or Dry Ice
Library Generation
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali