Learn More
Novus Biologicals™ Colon Matched Tumor and Normal Tissue Lysate
Codice prodotto. 18169898 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
2 Vials
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18169898 2 Vials pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18169898 Fornitore Novus Biologicals™ N. del fornitore NBP2470782VIAL
  • ONLINE EXCLUSIVE PRICE
    823.00€
    778.05€ / pezzi 2
    Risparmia 44.95€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 26-10-2026
Two matched 1 mg/mL vials (100 μg/vial) of human colon tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Colon Matched Tumor and Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 2 Vials
Content And Storage Store at -80C. Avoid freeze-thaw cycles.