Learn More
Shamrock™ Nastro per etichettatura multifunzione da 7,62 cm dotato di codice colore
Offered in a number of colors to fit your needs. Shamrock™ Color Coded Multi-Purpose 3 Inch Labeling Tape offers multiple opportunities for color coding in the laboratory - test, physician or department.
20.35€ - 55.95€
Specifications
Tipo di adesivo Removable
Product Type Color Coded Multi-Purpose Labeling Tape
Lunghezza (sistema metrico) 55 m
View More Specs
Products 29
Product Code Brand Colore Dimensions (LxW) Price Quantity & Availability  
Product Code Brand Colore Dimensions (LxW) Price Quantity & Availability  
11879080
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-120-26
Black 55 m x 13 mm
20.35€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11731636
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-6
Blue 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Add to basket
11827203
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-340-17
Chartreuse 55 m x 19 mm
27.50€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 22-05-2024
per visualizzare lo stock disponibile
Add to basket
11710095
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-100-23
Brown 55 m x 25 mm
35.45€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Add to basket
11770085
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-120-23
Brown 55 m x 13 mm
20.35€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11852592
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-100-10
Copper 55 m x 25 mm
35.45€
Pezzo
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Add to basket
11700095
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-340-23
Dark Brown 55 m x 19 mm
27.50€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Add to basket
11771616
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-110-29
Green, Fluorescent 55 m x 38 mm
48.15€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 22-05-2024
per visualizzare lo stock disponibile
Add to basket
11772255
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-27
Red, Fluorescent 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Add to basket
11790085
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-340-14
Grigio 55 m x 19 mm
27.50€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11831573
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-13
Lavender 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 22-05-2024
per visualizzare lo stock disponibile
Add to basket
11701636
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-3
Green 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Add to basket
11730454
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-100-5
Arancione 55 m x 25 mm
35.45€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11720085
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-120-14
Gray 55 m x 13 mm
20.35€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11897193
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-340-13
Lavender 55 m x 19 mm
27.50€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile
Add to basket
11721636
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-5
Orange 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11740414
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-120-7
Pink 55 m x 13 mm
20.35€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11750454
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-100-7
Pink 55 m x 25 mm
35.45€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11781616
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-110-4
Red 55 m x 38 mm
21.81€
12m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
16377205
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-110-4
Red 55 m x 38 mm
48.15€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Add to basket
11795753
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-120-4
Rosso 55 m x 24 mm
20.35€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 05-06-2024
per visualizzare lo stock disponibile
Add to basket
11711636
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-4
Red 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11710085
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-120-12
Violet 55 m x 13 mm
20.35€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11857153
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-100-12
Violet 55 m x 25 mm
35.45€
Pezzo
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 24-06-2024
per visualizzare lo stock disponibile
Add to basket
11862592
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-11
Rose 55 m x 50 mm
55.95€
Pezzo
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 13-06-2024
per visualizzare lo stock disponibile
Add to basket
11770434
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-100-1
Bianco 55 m x 25 mm
35.45€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Add to basket
11751616
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-110-1
White 55 m x 38 mm
48.15€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Add to basket
11791626
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-2
Yellow 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
11781626
missing translation for 'viewDocuments' Promotion Details
Shamrock™
ST-200-1
White 55 m x 50 mm
55.95€
55m
View alternative products
missing translation for 'toSaveUpto' 
»
In stock
per visualizzare lo stock disponibile
Add to basket
Description
Description
  • Resistant to water, oil and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
Specifications
Removable
Color Coded Multi-Purpose Labeling Tape
55 m
Videos
SDS
Documents
Documents
Certificates