Learn More
WTW™ Combined Fluoride Selective Electrode
Product Code. 10163482
Change view
Click to view available options
Connector Types:
DIN
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10163482 DIN Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10163482 Supplier WTW™ Supplier No. 106667
  • ONLINE EXCLUSIVE PRICE
    1365.00€
    1258.28€ / pezzi 1
    Risparmia 106.72€
    7% Sconto
View all promotions
In stock
The combined electrodes type 800 are excellent in their handling and offer the additional opportunity to also measure small sample volumes, as the ion selective and the reference electrodes are combined in a compact rod. Furthermore, they are convincing with their exceptionally great price/performance ratio.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Measuring small sample volumes
Connector Types DIN
Description Combined Fluoride Selective Electrode