Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1mm Thick
Sfoglia Tutto Fisherbrand ProdottiDescrizione
- Supplied as standard
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Sub-Gel Midi Horizontal Gel Electrophoresis Unit |
| Tipo | Comb |
| N. di pozzetti | 10 |
| Quantità | 1 Each |
| Spessore (sistema metrico) | 1 mm |
| Volume (sistema metrico) di campione | 27 μL |