missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA548869
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
COMMD9 Polyclonal antibody specifically detects COMMD9 in Human samples. It is validated for Western Blot.Specifica
COMMD9 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9P000 | |
COMMD9 | |
KLH conjugated synthetic peptide between 147-176 amino acids from the C-terminal region of human COMMD9. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
COMMD9 | |
1810029F08Rik; AA409293; COMM domain containing 9; COMM domain-containing protein 9; COMMD9; HSPC166; neuroprotective protein 9 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
29099 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |