Learn More
Emma Safety Footwear Comodius Safety Shoes
Product Code. 15988131
Change view
Click to view available options
Size:
39
40
41
42
43
44
46
47
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15988131 47 1 pair
15918131 39 1 pair
15928131 40 1 pair
15938131 41 1 pair
15948131 42 1 pair
15958131 43 1 pair
15968131 44 1 pair
15978131 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15988131 Supplier Emma Safety Footwear Supplier No. MM10207047
  • ONLINE EXCLUSIVE PRICE
    405.92€
    240.45€ / 1 paia
    Risparmia 165.47€
    40% Sconto
Spedizione stimata: 30-12-2026
High quality full grain leather upper and nappa leather lining. Exclusively developed for semi-orthopedic adjustments especially where customized in-soles are required.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Vibram™ Nitrile Rubber Heat Resistant Outsole
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Comodius
Liner Material Full grain Nappa leather
Material Leather
Size 47
Toe Material Aluminum