Learn More
BUCHI Condensate Trap for Rotavapor Condenser
Product Code. 15570570 Shop All Buchi Products
Change view
Click to view available options
Materiale:
Thermoplastic
Unit Size:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Materiale Unit Size
15570570 Thermoplastic Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15570570 Supplier BUCHI Supplier No. 11061985
Spedizione stimata: 30-12-2026
Collects and drains condensate, which may accumulate on the outside of the condenser.
  • For use with V (vertical coil) and HP (high-performance) condensers
  • Green
  • TPE
  • Drain hose connector diameter: 8 mm
TRUSTED_SUSTAINABILITY
Specifications
Materiale Thermoplastic
Tipo Condenser Insert