Learn More
Fisherbrand™ Condenser Recycle Water and Heat
A standard condenser that collects water in a tank rather than letting water out into a drain.
Marca:  Fisherbrand™
Dettagli aggiuntivi : Peso : 16.00000kg
Codice prodotto. 17117844
  • 6649.00€ / Pezzo
Spedizione stimata: 22-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Collects water in a tank
  • Collected water can then be used for washing in the next cycle
  • Heating element keeps temperature above 60°C to ensure that there is no possibility of bacteria developing in the tank
Specifica
Specifica
Condenser
Blue Line 6 / IQ6
60 Hz