Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, with GL Threads
Codice prodotto. 11943421
Cambia vista
Click to view available options
Nessun giunto rastremato standard:
NS 19/26
NS24/29
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Nessun giunto rastremato standard Dimensione della confezione
11943421 NS24/29 Pezzo
10233652 NS 19/26 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11943421 Fornitore Lenz Laborglasintrumente N. del fornitore 05223213
  • ONLINE EXCLUSIVE PRICE
    106.00€
    75.50€ / pezzi 1
    Risparmia 30.50€
    28% Sconto
Spedizione stimata: 25-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers acc. to DIN 12581 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifica
Lunghezza camicia (sistema metrico) 160 mm
Nessun giunto rastremato standard NS24/29