Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn With GL Threads
Codice prodotto. 11526804
Cambia vista
Click to view available options
Lunghezza camicia (sistema metrico):
500 mm
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Lunghezza camicia (sistema metrico) Dimensione della confezione
11526804 500 mm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11526804 Supplier Lenz Laborglasintrumente Supplier No. 05223514
  • ONLINE EXCLUSIVE PRICE
    155.00€
    103.81€ / pezzi 1
    Risparmia 51.19€
    33% Sconto
Spedizione stimata: 25-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Lunghezza camicia (sistema metrico) 500 mm
N. di lampadine 10
Nessun giunto rastremato standard NS 29/32