Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig, with GL Threads
Description
- Liebig condensers acc. to DIN 12576 and in addition to DIN
- For condensing vapours
- With NS socket and NS cone
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
Specifications
| Include | GL 14 Threads |
| Lunghezza (sistema metrico) | 250 mm |
| Nessun giunto rastremato standard | NS 14/23 |