Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Descrizione
- Condensers acc. to Liebig
- With olives Liebig condenser
- Acc. to DIN 12576
- For condensing vapours
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifica
Specifica
| Lunghezza (sistema metrico) | 400 mm |
| Nessun giunto rastremato standard | NS 14/23 |
| Materiale | Borosilicate Glass (3.3) |