Learn More
Lenz Laborglasintrumente Condensers acc. to Liebig, with Olives with Cone and Socket
Codice prodotto. 10664392
Cambia vista
Click to view available options
Nessun giunto rastremato standard:
Socket and Cone NS 29/32
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Nessun giunto rastremato standard Dimensione della confezione
10664392 Socket and Cone NS 29/32 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10664392 Fornitore Lenz Laborglasintrumente N. del fornitore 05210404
  • ONLINE EXCLUSIVE PRICE
    82.00€
    54.94€ / pezzi 1
    Risparmia 27.06€
    33% Sconto
Spedizione stimata: 23-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Liebig condenser acc. to DIN 12576
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifica
Lunghezza camicia (sistema metrico) 400 mm
Nessun giunto rastremato standard Socket and Cone NS 29/32