Learn More
Emma Safety Footwear Constans Safety Shoes
High quality full grain leather upper and nappa leather lining. Exclusively developed for semi-orthopedic adjustments especially where customized in-soles are required.
Marca: Emma Safety Footwear MM13207044
Dettagli aggiuntivi : Peso : 2.21000kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- Vibram™ Nitrile Rubber Heat Resistant Outsole
Specifica
EN ISO 20345:2011 | |
Black | |
Full grain Nappa leather | |
44 |
Laces | |
Constans | |
Leather | |
Aluminum |