Learn More
Bollé Safety Contour Polarized Safety Glasses
Product Code. 12699150 Shop All Bollé Safety Products
Product Code. 12699150 Supplier Bollé Safety Supplier No. CONTPOL
  • ONLINE EXCLUSIVE PRICE
    48.19€
    41.37€ / pezzi 1
    Risparmia 6.82€
    14% Sconto
Spedizione stimata: 10-12-2026
Protect workers with sturdy, protective, comfortable and lightweight safety spectacles. The Bolle Safety CONTOUR are designed to adapt perfectly to noses, so the frame sits perfectly on your face.
  • Lightweight and effective
  • Non-slip bridge
  • Non-slip TIPGRIP temples
  • Anti-static
  • Polarized lens offers unbeatable comfort by filtering glare and reflections
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Solar radiations risks: EN 172; Mechanical risk - Low energy impact - F
Lens Coating Scratch-resistant
Description Lightweight and effective safety spectacles
Frame Color Black
Weight (Metric) 21 g
Frame Type Adjustable Bridge