Learn More
Medline Scientific™ CETI 4.5kg Convex Refractometer
Product Code. 11965518
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11965518 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11965518 Supplier Medline Scientific™ Supplier No. 8200.0000M
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Spedizione stimata: 14-12-2026
Highly versatile refractometer that allows measurements of a wide range of substances. Medline Scientific™ CETI 4.5kg Convex Refractometer is capable of reading both reflected and transmitted light.
  • Wide range of measurable substances
  • Light entry ports for both upper and lower prisms
  • Supplied with a calibration set
    • Glass test piece
    • Alpha-Bromonaphtalene
    • Screwdriver
  • Supplied with dispersion tables, instruction manual, dust cover
Specifications
Type Convex Refractometer
Scale Range 0 to 95 Brix
Refractive Index 1.3000 to 1.7200
Resolution ±0.1°C
Includes 30x focusable telescope eyepiece, simultaneous scale reading, and border line setting
Quantity 1 Each
Weight (Metric) 4.5 kg
Dimensions (L x W x H) 280 x 190 x 380 mm
Height (Metric) 380 mm
Length (Metric) 280 mm
Temperature Range (Metric) 0°C to 200°C