Learn More
Epredia™ Cool-Cut with universal cassette clamp
Product Code. 11336770 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Standard Clamp
Universal Cassette Clamp
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11336770 Universal Cassette Clamp Each
12617886 Standard Clamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11336770 Supplier Epredia™ Supplier No. 771110
  • ONLINE EXCLUSIVE PRICE
    2565.00€
    2128.22€ / pezzi 1
    Risparmia 436.78€
    17% Sconto
In stock
Add to basket
Edge
Add to basket
Maintain a cool, consistent temperature for paraffin blocks during the sectioning process with the Epredia™ Cool-Cut accessory.
  • Directly fits onto the specimen cylinder of the rotary microtome line
  • Ability to be equipped as a universal cassette clamp or standard specimen clamp
  • Effective when extended cutting periods on the microtome are required to “step section” or “serial section” through the paraffin block
Includes:
  • Peltier device with integrated specimen clamp
  • Counter-balance weights
  • Power supply
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Cassette Clamp
For Use With (Equipment) Epredia Microtomes