missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407426
Click to view available options
Quantità:
100 g
500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]

Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Punti di ebollizione 1490°C
Materiale o nome chimico Copper(I) chloride
Punto di fusione 430°C
Forma fisica Powder
Quantità 100 g
Intervallo percentuale saggio 99% (Metal basis)
Informazioni di solubilità Very soluble in concentrated hydrochloric acid. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water.
Peso formulazione 98.99
Indice di rifrazione 1.93
Sensibilità Air and moisture sensitive
Aspetto Powder
Densità 4.14 g/cm3
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'