Learn More
Corning™ 96-Well Filter Plate Fluid Guard
Thermoformed polystyrene fluid guard
Marca: Corning™ 3514
Dettagli aggiuntivi : Peso : 0.77111kg Transport : UN number : 1684 Chem class : 6.1 Pack group : II
Descrizione
The fluid guard is manufactured from thermoformed polystyrene, non-sterile and easily snaps to the base of the filter plate. The unique design enables the guard to be applied manually or robotically to the plate. The guard serves to protect laboratory furniture and equipment from hazardous or radioactive materials.Specifica
Standard | |
Fluid Guard | |
96 | |
96-well filter plates | |
100/Pk. |
8 x 12 | |
Polystyrene | |
Non-sterile | |
100/Cs. |