Learn More
Corning™ Bovine Pituitary Extract (BPE)
Codice prodotto. 11533540 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
Quantità:
15 mg
75 mg
Dimensione della confezione:
15mg
75mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11533540 15 mg 15mg
11533620 75 mg 75mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11533540 Fornitore Corning™ N. del fornitore 354123
  • ONLINE EXCLUSIVE PRICE
    158.00€
    152.22€ / 15mg
    Risparmia 5.78€
    3% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 03-03-2027
Supplement used in the culture of a variety of epithelial and endothelial cells
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage 2° to 8°C
Product Type Bovine Pituitary Extract
Quantità 15 mg