Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Specifications
| Stile dei tubi | Heat sealable flexible tubing |
| Descrizione pellicola | EVA |
| N. per custodia | 1/Ea. |
| N. per confezione | 1/Pk. |
| Dettagli porta | Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site |
| Quantità | Each |
| Capacità (sistema metrico) | 1 L |