Learn More
Corning™ LSE™ Platform Rocker
Codice prodotto. 15995911 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
Modello:
Double level
Single level
Tipo di spina:
EU Plug
Tensione:
230 V
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Modello Tipo di spina Tensione Dimensione della confezione
15995911 Single level EU Plug 230 V Pezzo
15905921 Double level EU Plug 230 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15995911 Fornitore Corning™ N. del fornitore 6704
  • ONLINE EXCLUSIVE PRICE
    1025.00€
    954.93€ / pezzi 1
    Risparmia 70.07€
    6% Sconto
Spedizione stimata: 07-12-2026
Corning LSE Platform Rockers are ideal for Western Blotting, Gel staining and general mixing applications. Comes in single and double (30 x 30 cm) platform models to provide extra capacity.
The Corning™ LSE™ Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations. Both of these rugged models come with a 5-year warranty.
Features and Benefits
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
See resources below for sell sheet with specifications.
TRUSTED_SUSTAINABILITY
Specifica
Tensione 230 V
Product Type Platform Rocker
Tipo di spina EU Plug
Modello Single level