Learn More
Corning™ PF Media
BIOPHARMA
Codice prodotto. 15343701
Cambia vista
Click to view available options
Quantità:
6 x 500 mL
Dimensione della confezione:
pezzi 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15343701 6 x 500 mL pezzi 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15343701 Fornitore Corning™ N. del fornitore 40200CV
 Articolo soggetto a restrizioni
Spedizione stimata: 15-12-2026
Aggiungi al carrello
Aggiungi al carrello
Optimized for the cultivation of derivatives of the parental Chinese Hamster Ovary cell line, specifically CHO K-1
  • Proprietary serum-free and protein-free growth medium that does not contain any hormones or growth factors
  • Supports superior growth and viability over long-term pasasges in both adherent and suspension cell culture
  • Formulated without L-glutamine
For research use only. Not for IVD use.
TRUSTED_SUSTAINABILITY
Specifica
Linea cellulare CHO
Senza additivi No Glutamine
Quantità 6 x 500 mL