Learn More
Corning™ Reusable Gas Venting Cap for Vertical Sidearm Flasks
Codice prodotto. 10042132 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
For Use With: Vertical sidearm flasks, Fittings
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
10042132 For Use With: Vertical sidearm flasks, Fittings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10042132 Fornitore Corning™ N. del fornitore 4519177
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Used to provide gases into larger spinner flasks with vertical sidearms during cell culture
  • Used to provide gases into larger spinner flasks with vertical sidearms.
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • Gas filters are PTFE, 0.2 μm porosity.
  • The 316 stainless steel tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (apparecchiatura) Vertical sidearm flasks
N. per custodia 1/Cs.
N. per confezione 1/Pk.
Product Type Fittings