Learn More
Corning™ PuraMatrix™ Peptide Hydrogel
Codice prodotto. 10255252 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
Quantità:
5 mL
Dimensione della confezione:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
10255252 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10255252 Fornitore Corning™ N. del fornitore 354250
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Highly defined synthetic peptide assembles into fibrous 3D structure and promotes cell attachment
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Peptide hydrogel matrix
Quantità 5 mL