Descrizione
- Precise fit for 96 well Expanded Volume Polypropylene Microplate (Item #’s 3343 and 3344)
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
- Firm seal prevents evaporation and gas exchange
- Ideal for use in long-term, low temperature compound storage
Specifica
Specifica
| Sterilità | Non-sterile |
| No. of Wells | 96 |
| N. per custodia | 50/Cs. |
| N. per confezione | 10/Pk. |
| Product Type | Sealing Mat |
| Plate Compatibility | 96-well Polypropylene Plate |