Learn More
MedChemExpress Corticotropin-releasing factor (human)
Specifica
Specifica
| Purezza | 98.44% |
| CAS | 86784-80-7 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Da utilizzare con (applicazione) | Neuroscience-Neuromodulation |
| Peso molecolare | 4757.45 g/mol |
| Product Type | Peptides |
| Quantità | 5 mg |
| Shipping Condition | Room temperature |
| Solubilità | H2O : 25 mg/mL (5.25 mM∣ultrasonic and adjust pH to 2 with HCl) |
| Sinonimo | Human CRF∣ Human corticotropin-releasing factor |