Learn More
Corning™ Costar™ 384-Well Polypropylene Storage Block
Product Code. 10554581 Shop All Corning Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
10554581 Sterile Pack of 25
10266141 Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10554581 Supplier Corning™ Supplier No. 3964
  • ONLINE EXCLUSIVE PRICE
    373.00€
    348.48€ / pezzi 25
    Risparmia 24.52€
    6% Sconto
Spedizione stimata: 09-04-2027
Add to basket
Add to basket
Bulk Quote Request
Rigid sidewalls for optimal performance in automated handling
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polypropylene
No. of Wells 384
Sterility Sterile
Surface Treatment Untreated
Well Shape Round
For Use With (Application) For high throughput applications requiring added volume
No. per Case 25/Cs.
No. per Pack 5/Pk.
Quantity 25/Cs.
Well Color Clear