Learn More
Corning™ 8-Channel Adapter for Vacuum Aspirator
Product Code. 10327781 Shop All Corning Products
Change view
Click to view available options
Number of Channels:
8
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Number of Channels Unit Size
10327781 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10327781 Supplier Corning™ Supplier No. 4931
  • ONLINE EXCLUSIVE PRICE
    308.66€
    276.91€ / pezzi 1
    Risparmia 31.75€
    10% Sconto
In stock
Add to basket
Add to basket
Bulk Quote Request
For use with Corning Vacuum Aspirator. This 8-Channel Adapter can be used to remove media and solutions from 8 wells simultaneously on 96 well plate.
This is an 8-channel adapter that fits on the Aspirator (No. 4930) so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate. 
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Standard 1-200μL disposable pipet tips
Autoclavable Not Autoclavable
Number of Channels 8
Type Adapter, 8 channel
For Use With (Equipment) Corning Vacuum Aspirator (4930)