missing translation for 'onlineSavingsMsg'
Learn More

COX7A2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11416-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898473

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. COX7a2 (cytochrome c oxidase subunit VIIa polypeptide 2), also known as COX7AL or COX7AL1, is an 83 amino acid protein that localizes to the inner mitochondrial membrane and exists as a component of the COX complex, playing an important role in electron transport. COX7a2L (cytochrome c oxidase subunit 7A-related protein), also known as COX7AR or COX7RP, is an inner mitochondrial membrane protein that consists of 114 amino acids and is induced by estrogen.
Specifica

Specifica

COX7A2L
Polyclonal
Unconjugated
COX7A2L
COX7a related protein, COX7A2L, COX7AR, COX7RP, EB1, SIG81
Rabbit
Antigen Affinity Chromatography
RUO
20463, 298762, 9167
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14548, Q61387
COX7A2L
COX7A2L Fusion Protein Ag1980
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali