missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Giunti e raccordi a sgancio rapido serie MPC

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Marca:  CPC™ 22006T

Dettagli aggiuntivi : Peso : 0.00720kg

 vedi altre versioni di questo prodotto

Codice prodotto. 11960045

  • 28.00€ / pezzi 5


Spedizione stimata: 28-07-2021
Aggiungi al carrello

Descrizione

Descrizione

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 to 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Specifica

Specifica

USP Class VI, ADCF
160°F
60 psi
Autoclavable
ABS w/Silicone Seal USP Class VI
33 mm
9.5mm
-40°F
15.2 mm
White
71°C
Coupling Insert
9.5 mm
1.3 in.
Hose Barb Non-Valved In-line Coupling Insert
Cell culture media packaging and transfer lines, harvest and collection applications
-40°C
0.6 in.
Videos
SDS
Documenti
Certificati