Learn More
CPC Colder Products Company™ NS4 Series Quick-Disconnect Coupling Bodies
Product Code. 11900247
Change view
Click to view available options
Overall Length (Metric):
58 mm
59.5 mm
Product Type:
Hose Barb Valved In-line Coupling Body
NPT Valved Coupling Body
Unit Size:
Each
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Product Type unitSize
11900247 59.5 mm Hose Barb Valved In-line Coupling Body Each
11930247 58 mm NPT Valved Coupling Body Each
17493293 59.5 mm Hose Barb Valved In-line Coupling Body Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Product Code. 11900247 Supplier CPC Colder Products Company™ Supplier No. 3160323
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Manufactured from polypropylene. CPC Colder Products Company™ NS4 Series Quick-Disconnect Coupling Bodies are made from polypropylene. They have a temperature range between 0 ° to 71° and a pressure range up to 120 psi, 8.3 bar.
  • Polypropylene
  • -0°C to 71°C
  • Mounting: Thread
  • Pressure range: Vacuum to 120psi, 8.3 bar
  • Spilling: <0.10cc per disconnect, 0.10cc per connect
Specifications
Material EPDM
Overall Length (English) 2.34 in.
Overall Length (Metric) 59.5 mm
For Use With 6.4mm
Min. Operating Temperature (English) 32°F
Min. Operating Temperature (Metric) 0°C
Thread Size 0.25 in. hose barb
Height (Metric) 26.6 mm
Height (English) 1.05 in.
Includes Shut-off valves
Color Gray with Dark Gray Overmold
Maximum Pressure 120 psi
Max. Operating Temperature (English) 160°F
Max. Operating Temperature (Metric) 71°C
Product Type Hose Barb Valved In-line Coupling Body