Learn More
CPC Colder Products Company™ Giunti a sgancio rapido serie NS4
Codice prodotto. 17493293
Cambia vista
Click to view available options
Overall Length (Metric):
58 mm
59.5 mm
Product Type:
Corpo portagomma in linea con valvola
NPT Valved Coupling Body
Dimensione della confezione:
Pezzo
pezzi 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Overall Length (Metric) Product Type unitSize
17493293 59.5 mm Corpo portagomma in linea con valvola pezzi 25
11930247 58 mm NPT Valved Coupling Body Pezzo
11900247 59.5 mm Corpo portagomma in linea con valvola Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Codice prodotto. 17493293 Fornitore CPC Colder Products Company™ N. del fornitore NS4D17004
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
WARNING: Pressure, temperature, chemicals and operating environment can affect the performance of couplings. It is the customer’s responsibility to test the suitability of Colder products in their own application conditions.
Manufactured from polypropylene. CPC Colder Products Company™ NS4 Series Quick-Disconnect Coupling Bodies are made from polypropylene. They have a temperature range between 0 ° to 71° and a pressure range up to 120 psi, 8.3 bar.
  • Polypropylene
  • -0°C to 71°C
  • Mounting: Thread
  • Pressure range: Vacuum to 120psi, 8.3 bar
  • Spilling: <0.10cc per disconnect, 0.10cc per connect
Specifica
Materiale Polipropilene
Overall Length (English) 2.34 in.
Overall Length (Metric) 59.5 mm
Da utilizzare con 6.4mm
Min. Operating Temperature (English) 32°F
Min. Operating Temperature (Metric) 0°C
Dimensioni filo Tubo flessibile con flangia da 0.25 pollici
Altezza (sistema metrico) 26.6 mm
Altezza (sistema imperiale) 1.05 in.
Include Valvole d’arresto
Colore Grigio con rivestimento grigio scuro
Pressione massima 120 psi
Max. Operating Temperature (English) 160°F
Max. Operating Temperature (Metric) 71°C
Product Type Corpo portagomma in linea con valvola